2-(6-Methoxy-2-naphthyl)-2-methylpropanoic acid

Supplier: Matrix Scientific

054003-500MG 054003-2.5G
101905-804EA 364.62 USD
101905-804 101905-806
2-(6-Methoxy-2-naphthyl)-2-methylpropanoic acid
2-(6-Methoxy-2-naphthyl)-2-methylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₆O₃
MDL Number: MFCD01712996
CAS Number: 69337-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR