2-Methyl-2-(2-naphthyl)propanoic acid

Supplier: Matrix Scientific

054002-500MG 054002-2.5G
101905-802EA 341.21 USD
101905-802 101905-800
2-Methyl-2-(2-naphthyl)propanoic acid
2-Methyl-2-(2-naphthyl)propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₄O₂
MDL Number: MFCD11036968
CAS Number: 13365-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR