2-Methyl-2-(3-pyridinyl)propanoic acid

Supplier: Matrix Scientific

054000-500MG 054000-2.5G
101905-792EA 946.15 USD
101905-792 101905-794
2-Methyl-2-(3-pyridinyl)propanoic acid
2-Methyl-2-(3-pyridinyl)propanoic acid

Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD13563031
CAS Number: 169253-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR