2-(Cyclopentylmethoxy)-1-ethanol

Supplier: Matrix Scientific

053999-500MG 053999-2.5G
101905-790EA 838.46 USD
101905-790 101905-788
2-(Cyclopentylmethoxy)-1-ethanol
2-(Cyclopentylmethoxy)-1-ethanol

Hazard: Irritant

Formula: C₈H₁₆O₂
MDL Number: MFCD13563030
CAS Number: 1220019-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR