3-[2-(1-Pyrrolidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

053988-500MG 053988-2.5G
101905-752EA 364.62 USD
101905-752 101905-754
3-[2-(1-Pyrrolidinyl)ethyl]piperidine dihydrochloride
3-[2-(1-Pyrrolidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561634
CAS Number: 1220021-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR