2-(1-Ethyl-3-pyrrolidinyl)ethylamine

Supplier: Matrix Scientific

053979-500MG 053979-2.5G
101905-720EA 364.62 USD
101905-720 101905-722
2-(1-Ethyl-3-pyrrolidinyl)ethylamine
2-(1-Ethyl-3-pyrrolidinyl)ethylamine

Hazard: Irritant

Formula: C₈H₁₈N₂
MDL Number: MFCD09035754
CAS Number: 937670-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR