2-(1-Propyl-3-pyrrolidinyl)acetic acid

Supplier: Matrix Scientific

053974-500MG 053974-2.5G
101905-704EA 364.62 USD
101905-704 101905-706
2-(1-Propyl-3-pyrrolidinyl)acetic acid
2-(1-Propyl-3-pyrrolidinyl)acetic acid

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD13563019
CAS Number: 1220036-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR