2-(1-Ethyl-3-pyrrolidinyl)acetic acid

Supplier: Matrix Scientific

053972-500MG 053972-2.5G
101905-696EA 364.62 USD
101905-696 101905-698
2-(1-Ethyl-3-pyrrolidinyl)acetic acid
2-(1-Ethyl-3-pyrrolidinyl)acetic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD13563017
CAS Number: 1220027-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR