2-(3-Pyrrolidinylidene)acetic acid

Supplier: Matrix Scientific

053968-2.5G 053968-500MG
101905-680EA 364.62 USD
101905-680 101905-682
2-(3-Pyrrolidinylidene)acetic acid
2-(3-Pyrrolidinylidene)acetic acid

Hazard: Irritant

Formula: C₆H₉NO₂
MDL Number: MFCD13563014
CAS Number: 5626-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR