2-(1-Methyl-3-piperidinyl)acetic acid

Supplier: Matrix Scientific

053960-500MG 053960-2.5G
101905-648EA 384.62 USD
101905-648 101905-650
2-(1-Methyl-3-piperidinyl)acetic acid
2-(1-Methyl-3-piperidinyl)acetic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD11554179
CAS Number: 169253-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR