4-(2-Fluorobenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053953-500MG 053953-2.5G
101905-620EA 384.62 USD
101905-620 101905-622
4-(2-Fluorobenzyl)-1,4-diazepan-5-one
4-(2-Fluorobenzyl)-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₂H₁₅FN₂O
MDL Number: MFCD13563005
CAS Number: 1220030-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR