4-(3-Fluorobenzyl)-1,4-diazepan-5-one

Supplier: Matrix Scientific

053949-500MG 053949-2.5G
101905-604EA 364.62 USD
101905-604 101905-606
4-(3-Fluorobenzyl)-1,4-diazepan-5-one
4-(3-Fluorobenzyl)-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₁₂H₁₅FN₂O
MDL Number: MFCD07791560
CAS Number: 879050-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR