4-Butyl-1,4-diazepan-5-one

Supplier: Matrix Scientific

053944-500MG 053944-2.5G
101905-584EA 364.62 USD
101905-584 101905-586
4-Butyl-1,4-diazepan-5-one
4-Butyl-1,4-diazepan-5-one

Hazard: Irritant

Formula: C₉H₁₈N₂O
MDL Number: MFCD13562998
CAS Number: 1220027-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR