4-(2-Methoxyethyl)-1,4-diazepan-5-one hydrochloride

Supplier: Matrix Scientific

053942-500MG 053942-2.5G
101905-576EA 364.62 USD
101905-576 101905-578
4-(2-Methoxyethyl)-1,4-diazepan-5-one hydrochloride
4-(2-Methoxyethyl)-1,4-diazepan-5-one hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD27579023
CAS Number: 1219960-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR