2-Amino-1-(3-hydroxy-1-azetidinyl)-3-phenyl-1-propanone

Supplier: Matrix Scientific

053927-500MG 053927-2.5G
101905-520EA 364.62 USD
101905-520 101905-522
2-Amino-1-(3-hydroxy-1-azetidinyl)-3-phenyl-1-propanone
2-Amino-1-(3-hydroxy-1-azetidinyl)-3-phenyl-1-propanone

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD13562990
CAS Number: 1236264-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR