N-(3-Azetidinyl)benzamide
Supplier: Matrix Scientific
053922-2.5G
053922-500MG
101905-500EA
653.85
USD
101905-500
101905-502
N-(3-Azetidinyl)benzamide
N-(3-Azetidinyl)benzamide
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O |
MDL Number:
MFCD13562986 CAS Number: 1219979-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...