4-(3-Azetidinyl)morpholine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(Azetidin-3-yl)morpholine dihydrochloride

053921-500MG 053921-2.5G
101905-496EA 384.62 USD
101905-496 101905-498
4-(3-Azetidinyl)morpholine dihydrochloride
4-(3-Azetidinyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₆Cl₂N₂O
MW: 215.12 g/mol
MDL Number: MFCD11617823
CAS Number: 178312-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR