2-[(Benzyloxy)methyl]azetidine

Supplier: Matrix Scientific

053910-500MG 053910-2.5G
101905-452EA 2238.46 USD
101905-452 101905-454
2-[(Benzyloxy)methyl]azetidine
2-[(Benzyloxy)methyl]azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD13562980
CAS Number: 1220030-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR