3-[(Benzyloxy)methyl]azetidine

053906-2.5G
101905-438EA 838.46 USD
101905-438
3-[(Benzyloxy)methyl]azetidine
3-[(Benzyloxy)methyl]azetidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD13562976
CAS Number: 1220038-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR