(1-Benzyl-3-azetidinyl)methanamine
Supplier: Matrix Scientific
053901-2.5G
053901-500MG
101905-416EA
364.62
USD
101905-416
101905-418
(1-Benzyl-3-azetidinyl)methanamine
(1-Benzyl-3-azetidinyl)methanamine
Hazard: Irritant
Formula:
C₁₁H₁₆N₂ |
MDL Number:
MFCD13562971 CAS Number: 1219967-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...