2-(1-Methyl-3-oxo-2-piperazinyl)acetic acid
Supplier: Matrix Scientific
053899-2.5G
053899-500MG
101905-408EA
364.62
USD
101905-408
101905-410
2-(1-Methyl-3-oxo-2-piperazinyl)acetic acid
2-(1-Methyl-3-oxo-2-piperazinyl)acetic acid
Hazard: Irritant
Formula:
C₇H₁₂N₂O₃ |
MDL Number:
MFCD05870556 CAS Number: 1033537-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...