2-(1-Isopropyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Supplier: Matrix Scientific

053896-500MG 053896-2.5G
101905-396EA 364.62 USD
101905-396 101905-398
2-(1-Isopropyl-4-methyl-3-oxo-2-piperazinyl)acetic acid
2-(1-Isopropyl-4-methyl-3-oxo-2-piperazinyl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD13562969
CAS Number: 1236267-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR