Methyl-2-(4-methyl-3-oxo-2-piperazinyl)acetate
Supplier: Matrix Scientific
053891-2.5G
053891-500MG
101905-376EA
364.62
USD
101905-376
101905-378
Methyl-2-(4-methyl-3-oxo-2-piperazinyl)acetate
Methyl-2-(4-methyl-3-oxo-2-piperazinyl)acetate
Hazard: Irritant
Formula:
C₈H₁₄N₂O₃ |
MDL Number:
MFCD13562965 CAS Number: 534603-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...