2-Methoxy-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol

Supplier: Matrix Scientific

053877-500MG 053877-2.5G
101905-320EA 364.62 USD
101905-320 101905-322
2-Methoxy-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol
2-Methoxy-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol

Hazard: Irritant

Formula: C₇H₉N₃O₂
MW: 167.17 g/mol
MDL Number: MFCD13562956
CAS Number: 1220030-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR