2-Phenyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride

Supplier: Matrix Scientific

053873-2.5G 053873-500MG
101905-306EA 838.46 USD
101905-306 101905-304
2-Phenyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride
2-Phenyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃Cl₂N₃
MDL Number: MFCD16615182
CAS Number: 1380571-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR