2-(tert-Butyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine
Supplier: Matrix Scientific
053871-2.5G
053871-500MG
101905-300EA
364.62
USD
101905-300
101905-302
2-(tert-Butyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine
2-(tert-Butyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine
Hazard: Irritant
Formula:
C₁₀H₁₅N₃ MW: 177.25 g/mol |
MDL Number:
MFCD13562952 CAS Number: 1211584-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...