4-(6-Chloro-4-pyrimidinyl)-2-piperazinone
Supplier: Matrix Scientific
051845-2.5G
051845-500MG
101897-274EA
838.46
USD
101897-274
101897-272
4-(6-Chloro-4-pyrimidinyl)-2-piperazinone
4-(6-Chloro-4-pyrimidinyl)-2-piperazinone
Hazard: Irritant
Formula:
C₈H₉ClN₄O |
MDL Number:
MFCD13561823 CAS Number: 1220020-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...