1-(6-Chloro-4-pyrimidinyl)azepane
Supplier: Matrix Scientific
051844-2.5G
051844-500MG
101897-268EA
364.62
USD
101897-268
101897-270
1-(6-Chloro-4-pyrimidinyl)azepane
1-(6-Chloro-4-pyrimidinyl)azepane
Hazard: Irritant
Formula:
C₁₀H₁₄ClN₃ |
MDL Number:
MFCD13561822 CAS Number: 1220033-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...