N,N-Diallyl-6-chloro-4-pyrimidinamine
Supplier: Matrix Scientific
051838-500MG
051838-2.5G
101897-246EA
838.46
USD
101897-246
101897-244
N,N-Diallyl-6-chloro-4-pyrimidinamine
N,N-Diallyl-6-chloro-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₀H₁₂ClN₃ |
MDL Number:
MFCD13561818 CAS Number: 1220036-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...