6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinylamine
Supplier: Matrix Scientific
051825-500MG
051825-2.5G
101897-194EA
838.46
USD
101897-194
101897-192
6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinylamine
6-(2,3-Dihydro-1H-indol-1-yl)-3-pyridinylamine
Hazard: Irritant
Formula:
C₁₃H₁₃N₃ |
MDL Number:
MFCD08700223 CAS Number: 937603-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...