4-(5-Amino-2-pyridinyl)-2-piperazinone
Supplier: Matrix Scientific
051816-500MG
051816-2.5G
101897-158EA
838.46
USD
101897-158
101897-156
4-(5-Amino-2-pyridinyl)-2-piperazinone
4-(5-Amino-2-pyridinyl)-2-piperazinone
Hazard: Irritant
Formula:
C₉H₁₂N₄O |
MDL Number:
MFCD09040668 CAS Number: 926262-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...