1-(5-Amino-2-pyridinyl)-3-pyrrolidinol
Supplier: Matrix Scientific
051807-2.5G
051807-500MG
101897-122EA
838.46
USD
101897-122
101897-120
1-(5-Amino-2-pyridinyl)-3-pyrrolidinol
1-(5-Amino-2-pyridinyl)-3-pyrrolidinol
Hazard: Irritant
Formula:
C₉H₁₃N₃O |
MDL Number:
MFCD11849347 CAS Number: 939376-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...