N-Benzyl-5-bromo-N-ethyl-4-methyl-2-pyridinamine

Supplier: Matrix Scientific

051800-500MG
101897-092EA 272.55 USD
101897-092
N-Benzyl-5-bromo-N-ethyl-4-methyl-2-pyridinamine
N-Benzyl-5-bromo-N-ethyl-4-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₅H₁₇BrN₂
MDL Number: MFCD13561812
CAS Number: 1220019-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR