N-Benzyl-5-bromo-N,4-dimethyl-2-pyridinamine

051794-2.5G
101897-070EA 1294.68 USD
101897-070
N-Benzyl-5-bromo-N,4-dimethyl-2-pyridinamine
N-Benzyl-5-bromo-N,4-dimethyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₄H₁₅BrN₂
MDL Number: MFCD13561806
CAS Number: 1219960-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR