5-Bromo-4-methyl-N,N-dipropyl-2-pyridinamine
Supplier: Matrix Scientific
051791-2.5G
051791-500MG
101897-058EA
838.46
USD
101897-058
101897-056
5-Bromo-4-methyl-N,N-dipropyl-2-pyridinamine
5-Bromo-4-methyl-N,N-dipropyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₂H₁₉BrN₂ |
MDL Number:
MFCD13561803 CAS Number: 1219964-53-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...