5-Bromo-4-methyl-2-(4-methyl-1-piperidinyl)pyridine
Supplier: Matrix Scientific
051785-2.5G
051785-500MG
101897-034EA
838.46
USD
101897-034
101897-032
5-Bromo-4-methyl-2-(4-methyl-1-piperidinyl)pyridine
5-Bromo-4-methyl-2-(4-methyl-1-piperidinyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₁₇BrN₂ MW: 269.19 g/mol |
MDL Number:
MFCD13561798 CAS Number: 1219964-45-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...