4-(Dipropylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

051762-500MG 051762-2.5G
101896-942EA 838.46 USD
101896-942 101896-940
4-(Dipropylamino)-3-nitrobenzoic acid
4-(Dipropylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD12445716
CAS Number: 960060-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR