4-[(2-Hydroxyethyl)(methyl)amino]-3-nitrobenzoic acid
Supplier: Matrix Scientific
051753-2.5G
051753-500MG
101896-906EA
838.46
USD
101896-906
101896-904
4-[(2-Hydroxyethyl)(methyl)amino]-3-nitrobenzoic acid
4-[(2-Hydroxyethyl)(methyl)amino]-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₅ |
MDL Number:
MFCD13205895 CAS Number: 1183788-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...