N-Methyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051712-2.5G 051712-500MG
101896-740EA 364.62 USD
101896-740 101896-742
N-Methyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride
N-Methyl-N-(2-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561764
CAS Number: 1219964-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR