4-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

051705-500MG 051705-2.5G
101896-712EA 364.62 USD
101896-712 101896-714
4-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride
4-Methyl-1-(2-pyrrolidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561757
CAS Number: 1220027-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR