4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride

Supplier: Matrix Scientific

051698-500MG 051698-2.5G
101896-684EA 364.62 USD
101896-684 101896-686
4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MDL Number: MFCD13561750
CAS Number: 1220035-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR