1-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051690-500MG 051690-2.5G
101896-652EA 364.62 USD
101896-652 101896-654
1-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride
1-(2-Piperidinylmethyl)-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂
MDL Number: MFCD13561742
CAS Number: 1016843-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR