2-[4-(2-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

051687-2.5G
101896-642EA 838.46 USD
101896-642
2-[4-(2-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride
2-[4-(2-Piperidinylmethyl)-1-piperazinyl]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₇Cl₂N₃O
MDL Number: MFCD13561739
CAS Number: 1220018-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR