N-Methyl(phenyl)-N-(2-piperidinylmethyl)methanamine dihydrochloride

051685-2.5G
101896-634EA 838.46 USD
101896-634
N-Methyl(phenyl)-N-(2-piperidinylmethyl)methanamine dihydrochloride
N-Methyl(phenyl)-N-(2-piperidinylmethyl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₄Cl₂N₂
MW: 291.27 g/mol
MDL Number: MFCD13561737
CAS Number: 1220019-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR