N-Methyl-N-(2-piperidinylmethyl)cyclohexanamine dihydrochloride

051683-2.5G
101896-626EA 1061.78 USD
101896-626
N-Methyl-N-(2-piperidinylmethyl)cyclohexanamine dihydrochloride
N-Methyl-N-(2-piperidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561735
CAS Number: 1219960-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR