N-(2-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride

051682-2.5G
101896-622EA 1339.89 USD
101896-622
N-(2-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride
N-(2-Piperidinylmethyl)-N-propyl-1-propanamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₈Cl₂N₂
MDL Number: MFCD13561734
CAS Number: 1220018-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR