1-(2-Piperidinylmethyl)-4-piperidinol dihydrochloride

051681-2.5G
101896-618EA 838.46 USD
101896-618
1-(2-Piperidinylmethyl)-4-piperidinol dihydrochloride
1-(2-Piperidinylmethyl)-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂O
MDL Number: MFCD13561733
CAS Number: 1219964-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR