4-(2-Piperidinylmethyl)-2-piperazinone dihydrochloride

051680-2.5G
101896-614EA 838.46 USD
101896-614
4-(2-Piperidinylmethyl)-2-piperazinone dihydrochloride
4-(2-Piperidinylmethyl)-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁Cl₂N₃O
MDL Number: MFCD13561732
CAS Number: 1219963-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR