1-Benzyl-4-(2-piperidinylmethyl)piperazine dihydrochloride

051678-2.5G
101896-606EA 838.46 USD
101896-606
1-Benzyl-4-(2-piperidinylmethyl)piperazine dihydrochloride
1-Benzyl-4-(2-piperidinylmethyl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₇H₂₉Cl₂N₃
MDL Number: MFCD13561730
CAS Number: 1220021-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR