4-Methyl-1-(2-piperidinylmethyl)piperidine dihydrochloride

051676-2.5G
101896-598EA 838.46 USD
101896-598
4-Methyl-1-(2-piperidinylmethyl)piperidine dihydrochloride
4-Methyl-1-(2-piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561728
CAS Number: 1220018-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR